Order Entry
Slovenia
ContactUsLinkComponent
Adaptor-unf for prominent pumps, unf1/4 m 20 x 1,5 ø 0,8 mm, ptfe reinforced with glass fibres, 1x female thread m20 x 1, 1x femal e thread unf 1/4 28g, bore dia. 0,8 mm 1 * 1 items
  BOHLD731-12
 :  Bohlender
undefined
Adaptor-unf for prominent pumps, unf1/4 m 20 x 1,5 ø 0,8 mm, ptfe reinforced with glass fibres, 1x female thread m20 x 1, 1x femal e thread unf 1/4 28g, bore dia. 0,8 mm 1 * 1 items
  BOHLD731-12
 :  Bohlender
Frequently Bought Together