Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Computers for Laboratory Equipment
Dell™ mouse, bundled with Dell™ keyboard, ES
Catalog # 906-0218
Computers for Laboratory Equipment
Catalog # 906-0218
Computers for Laboratory Equipment
Computers for Laboratory Equipment
Dell™ mouse, bundled with Dell™ keyboard, ES
Price
30,60 €
Log in for availability
1 * 1 items
Product Details & Documents
PC equipment optimised for use with laboratory devices and software. Designed to meet laboratory conditions, ensuring necessary performance. Includes a wide range of accessories such as monitors and keyboards.
  • Pre-installed with Windows LTSC 10 or 11 to reduce downtime from frequent updates and increase stability
  • Compact footprint with integrated PC-monitor solutions
  • Pre-installed language packs
  • Keyboards with all common European language layouts
  • Compatible with main CDS (Chromatography Data Systems)
Specifications
  • Description:
    Dell™ mouse, bundled with Dell™ keyboard, ES
  • Seq:
    23
Complementary Products
Product Information
  • Catalog #
    Description
    Availability
    Price
  • 906-0219
    Dell™ mouse, bundled with Dell™ keyboard, DE
    Log in for availability
    1 * 1 items - 31,60 €
  • 906-0211
    Dell™ mouse, bundled with Dell™ keyboard, FR
    Log in for availability
    1 * 1 items - 31,60 €