Order Entry
Slovenia
ContactUsLinkComponent
Neptune 1000 µl pipette tips. bulk, 1 bag of 1000 tips/pack, 4 bags/case. proprietary sample saving surface (s3) technology eliminat es sample hold-up providing accurate sample delivery. universal fit ideal for both single and multichannel pipettors 1 * 1.000 items
Catalog # BITX2370
Supplier:  BIOTIX
Neptune 1000 µl pipette tips. bulk, 1 bag of 1000 tips/pack, 4 bags/case. proprietary sample saving surface (s3) technology eliminat es sample hold-up providing accurate sample delivery. universal fit ideal for both single and multichannel pipettors 1 * 1.000 items
Catalog # BITX2370
Supplier:  BIOTIX
Frequently Bought Together