Order Entry
Slovenia
ContactUsLinkComponent
Elite-bac advantage columns are exceptionally robust, ensuring longer column lifetime and highly reproducible data for analysis of b lood alcohol content (bac).inner diameter (mm i.d): 0.53length (m): 30film thickness (?m): 3.00. key benefits include 1 * 1 items
Catalog # PERCN9315072
Supplier:  PerkinElmer
Elite-bac advantage columns are exceptionally robust, ensuring longer column lifetime and highly reproducible data for analysis of b lood alcohol content (bac).inner diameter (mm i.d): 0.53length (m): 30film thickness (?m): 3.00. key benefits include 1 * 1 items
Catalog # PERCN9315072
Supplier:  PerkinElmer
Frequently Bought Together