Order Entry
Slovenia
ContactUsLinkComponent
Elite-bac advantage columns are exceptionally robust, ensuring longer column lifetime and highly reproducible data for analysis of b lood alcohol content (bac). inner diameter (mm i.d.): 0.32length (m): 30film thickness (?m): 1.80. key benefits inclu 1 * 1 items
Catalog # PERCN9315071
Supplier:  PerkinElmer
Elite-bac advantage columns are exceptionally robust, ensuring longer column lifetime and highly reproducible data for analysis of b lood alcohol content (bac). inner diameter (mm i.d.): 0.32length (m): 30film thickness (?m): 1.80. key benefits inclu 1 * 1 items
Catalog # PERCN9315071
Supplier:  PerkinElmer
Frequently Bought Together