Order Entry
Slovenia
ContactUsLinkComponent
- Safety dispensing lid effectively prevents contamination - Safely restrains dripping residue - For 205 litre steel bung hole drums - Openings for 3/4 and 2 bung hole - for using pumps and/or fill-level indicators - Grooved storage area ensures di 1 * 1 items
Catalog # MSGA13303
Supplier:  asecos
- Safety dispensing lid effectively prevents contamination - Safely restrains dripping residue - For 205 litre steel bung hole drums - Openings for 3/4 and 2 bung hole - for using pumps and/or fill-level indicators - Grooved storage area ensures di 1 * 1 items
Catalog # MSGA13303
Supplier:  asecos
Frequently Bought Together