3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 99+%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 99+%
Catalog # J60332.06
Supplier: Thermo Fisher Scientific
CAS Number: 207738-08-7
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 207738-08-7 Molecular Formula: C16H20N2.2HCl.xH2O Formula Weight: 313.27(anhy) Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Moisture and Light Sensitive Appearance: White to off-white MDL No.: MFCD00150104
Excellent Colourimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
Synonyms: 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine dihydrochloride xhydrate, TMB-HCl hydrate, Dihydrochloride TMB hydrate
- CAS Number:207738-08-7
- Formula:C₁₆H₂₀N₂·2HCl·xH₂O
- MDL Number:MFCD00150104
- Molecular Weight:313.27 g/mol (anhy)
- Storage Temperature:Freezer
- EINECS:264-769-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



