3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 99+%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 99+%
Catalog # J60332.03
Supplier: Thermo Fisher Scientific
CAS Number: 207738-08-7
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 207738-08-7 Molecular Formula: C16H20N2.2HCl.xH2O Formula Weight: 313.27(anhy) Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Moisture and Light Sensitive Appearance: White to off-white MDL No.: MFCD00150104
Excellent Colourimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
Synonyms: 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine dihydrochloride xhydrate, TMB-HCl hydrate, Dihydrochloride TMB hydrate
- CAS Number:207738-08-7
- Formula:C₁₆H₂₀N₂·2HCl·xH₂O
- MDL Number:MFCD00150104
- Molecular Weight:313.27 g/mol (anhy)
- Storage Temperature:Freezer
- EINECS:264-769-6
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



