Order Entry
Slovenia
ContactUsLinkComponent
Energy efficient motor rated less than 1 amp UV light fixture on a push button timer with door interlock Fluorescent Light fixture UV resisitant clear plexiglas closing face shield 1 * 1 ST
Catalog # IBSAAU-110-HOOD-HE
Supplier:  IBS Stock Items
Energy efficient motor rated less than 1 amp UV light fixture on a push button timer with door interlock Fluorescent Light fixture UV resisitant clear plexiglas closing face shield 1 * 1 ST
Catalog # IBSAAU-110-HOOD-HE
Supplier:  IBS Stock Items
Frequently Bought Together