Order Entry
Slovenia
ContactUsLinkComponent
Magnesia 16 Automated NA Isolation System Method: Full Automated Isolation with Cellulose Coated Magnetic Beads Capacity: 1-16 Samples Sample Volume: 200 / 400 / 1200 µl Elution Volume: 60 / 100 / 150 / 200 µl Efficiency: 6 mg / 200 µl Heating Block: 1 * 1 ST
Catalog # IBSADE-010
Supplier:  IBS Stock Items
Magnesia 16 Automated NA Isolation System Method: Full Automated Isolation with Cellulose Coated Magnetic Beads Capacity: 1-16 Samples Sample Volume: 200 / 400 / 1200 µl Elution Volume: 60 / 100 / 150 / 200 µl Efficiency: 6 mg / 200 µl Heating Block: 1 * 1 ST
Catalog # IBSADE-010
Supplier:  IBS Stock Items
Frequently Bought Together