Order Entry
Slovenia
ContactUsLinkComponent
Sucrose Standard 0% - 1.332986 (± 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals pla ce. Extended shelf-life of 12 weeks 1 * 15 mL
Catalog # REGEBS00
Supplier:  REAGECON
Sucrose Standard 0% - 1.332986 (± 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals pla ce. Extended shelf-life of 12 weeks 1 * 15 mL
Catalog # REGEBS00
Supplier:  REAGECON
Frequently Bought Together