Order Entry
Slovenia
ContactUsLinkComponent
Noise-insulated vacuum pump module with built-in pump B04134 and energy-saving control, antivibration metal plate, full-sight door g lass, and connecting accessories for VO500 1 * 1 items
Catalog # 466-0415
Supplier:  MEMMERT
Noise-insulated vacuum pump module with built-in pump B04134 and energy-saving control, antivibration metal plate, full-sight door g lass, and connecting accessories for VO500 1 * 1 items
Catalog # 466-0415
Supplier:  MEMMERT
Frequently Bought Together