Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Electrode cleaning solutions

Electrode cleaning solutions

Pepsin-HCl solution, for cleaning junctions with protein contamination, treatment time about 1 h, 1×250 ml, 250 mL
Catalog # METN51340068
Supplier:   Mettler - Toledo
Electrode cleaning solutions
Catalog # METN51340068
Supplier:  Mettler - Toledo
CAS Number:  
Electrode cleaning solutions

Pepsin-HCl solution, for cleaning junctions with protein contamination, treatment time about 1 h, 1×250 ml, 250 mL

CAS Number:  
Price
33,16 €
Log in for availability
1 * 250 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Pepsin-HCl solution, for cleaning junctions with protein contamination, treatment time about 1 h, 1×250 ml

Specifications