Pipette tips
Product Details & Documents
These pipette tips are made from virgin PP or exclusive S3 polymer.
- Polymer tips retain virtually no fluids, allowing the user to dispense truly accurate sample volumes
- Environmentally friendly packaging: some tips available in ESP reload systems
- Tips are certified free of RNase, DNase, DNA and endotoxins
- Graduated sides
The exclusive S3 polymer delivers to within 0,1% of indicated volume, eliminating sample hold-up and ensuring a more precise application. This proprietary technology is essential for quantitative assays where sample retention can hinder accurate results. Universal pipette tips are made from virgin PP and are equipped with a thin wall design that provides an optimal seal with industry leading pipettors.
Eco-friendly empty pipette tip trays are manufactured from recycled PP. The trays are autoclavable, enabling reuse for maximum sustainability. When combined with ESP reload systems, these tip trays reduce plastic waste by up to 90%. The trays provide superior stability, firm placement, and a maximum space saving design.
Specifications
- Packed:Bulk, 1000 tips/bag
- Sterile:-
- Volume:200 µl
- Length:50,75 mm
- Tip style:Universal, S³ polymer
- Tip colour:Transparent



