Order Entry
Slovenia
ContactUsLinkComponent
KIT, PROSTAGLANDIN E2 ELISA KIT, PGE2 (FPIA) ELISA KIT IS A COMPLETE KIT FOR THE QUANTITATIVE DETERMINATION OF PROSTAGLANDIN E2 IN B UFFERS AND CULTURE MEDIA. 1 * 1 KIT
Catalog # ABNOKA0325
Supplier:  Abnova
KIT, PROSTAGLANDIN E2 ELISA KIT, PGE2 (FPIA) ELISA KIT IS A COMPLETE KIT FOR THE QUANTITATIVE DETERMINATION OF PROSTAGLANDIN E2 IN B UFFERS AND CULTURE MEDIA. 1 * 1 KIT
Catalog # ABNOKA0325
Supplier:  Abnova