Order Entry
Slovenia
ContactUsLinkComponent
KIT, METANEPHRINE/NORMETANEPHRINE ELISA KIT, METANEPHRINE/NORMETANEPHRINE ELISA KIT IS AN ENZYME IMMUNOASSAY FOR THE QUANTITATIVE DE TERMINATION OF FREE METANEPHRINE AND FREE NORMETANEPHRINE IN PLASMA. 1 * 1 KIT
Catalog # ABNOKA1878
Supplier:  Abnova
KIT, METANEPHRINE/NORMETANEPHRINE ELISA KIT, METANEPHRINE/NORMETANEPHRINE ELISA KIT IS AN ENZYME IMMUNOASSAY FOR THE QUANTITATIVE DE TERMINATION OF FREE METANEPHRINE AND FREE NORMETANEPHRINE IN PLASMA. 1 * 1 KIT
Catalog # ABNOKA1878
Supplier:  Abnova