Order Entry
Slovenia
ContactUsLinkComponent
Freezer, gnp2666 upright, premium nofrost, gross capacity, freezer compartment [l] 265 energy efficiency class* a+++ energy consumpt ion in 3 65 days* [kwh/a] 146, door hinges right interchangeable 1 * 1 items
  LIEBGNP2666
 :  Liebherr
Freezer, gnp2666 upright, premium nofrost, gross capacity, freezer compartment [l] 265 energy efficiency class* a+++ energy consumpt ion in 3 65 days* [kwh/a] 146, door hinges right interchangeable 1 * 1 items
  LIEBGNP2666
 :  Liebherr
Frequently Bought Together