LRC columns offer 4 internal diameters and various lengths to provide a range of bed heights and column volumes. Made of borosilicat e glass and equipped with one adjustable and one fixed piston. LRC COLUMN 15MM/80-200MM, LRC15X080-200V01 1 * 1 items
Catalog # 554-2372
Supplier: Cytiva (Formerly Pall Lab)
Frequently Bought Together