LRC columns offer 4 internal diameters and various lengths to provide a range of bed heights and column volumes. Made of borosilicat e glass and equipped with one adjustable and one fixed piston. LRC COLUMN 10MM/330-450MM, LRC10X330-450V01 1 * 1 items
Catalog # 554-2365
Supplier: Cytiva (Formerly Pall Lab)
Frequently Bought Together