Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Spanner wrenches
Spanner wrenches
Spanner wrench, for nuts Ø 65 - 75 mm, for HiScale™ 26 columns, Galvanised steel
Catalog # 28-9647-77
Supplier:   Cytiva
Spanner wrenches
Catalog # 28-9647-77
Supplier:  Cytiva
Spanner wrenches
Spanner wrenches
Spanner wrench, for nuts Ø 65 - 75 mm, for HiScale™ 26 columns, Galvanised steel
Price
96,10 €
Log in for availability
1 * 2 items
Product Details & Documents
These spanner wrenches are used with HiScale™ columns. To readjust the position of the adapter in the column a spanner can be used to turn the end cap. To prevent the column from rotating, a second spanner can be used to hold the end housing.
Specifications
  • Description:
    Spanner wrench, for nuts Ø 65 - 75 mm, for HiScale™ 26 columns
  • Material:
    Galvanised steel
  • Seq:
    02