Order Entry
Slovenia
ContactUsLinkComponent
SOLUTION DE LIMITES DE QUANTITIFICATION ELEMENTS MAJEURS 5 components Ca 600mg/l Mg 600mg/l Na 180mg/l K 600mg/l S 400mg/l in HNO3 5% 1 * 125 mL
Catalog # CMPLA5B4.K6.5N
SOLUTION DE LIMITES DE QUANTITIFICATION ELEMENTS MAJEURS 5 components Ca 600mg/l Mg 600mg/l Na 180mg/l K 600mg/l S 400mg/l in HNO3 5% 1 * 125 mL
Catalog # CMPLA5B4.K6.5N
Frequently Bought Together