Spanner wrenches
Spanner wrench, for nuts Ø 95 - 100 mm, for HiScale™ 50 columns, Galvanised steel
28-9647-78
: Cytiva
These spanner wrenches are used with HiScale™ columns. To readjust the position of the adapter in the column a spanner can be used to turn the end cap. To prevent the column from rotating, a second spanner can be used to hold the end housing.
- Description:Spanner wrench, for nuts Ø 95 - 100 mm, for HiScale™ 50 columns
- Material:Galvanised steel
- Seq:03