Disinfectant solutions, Bacillol AF
Product Details & Documents
Alcohol based disinfectant, aldehyde-free and fragrance-free, suitable for spray wipe of all surfaces resistant to alcohol in the agro-food and pharmaceutical industries, in hospital environments or in medical practices. Suitable for application where rapid disinfection is required that leaves no residue.
Contents:
Propan-1-ol 450 mg/g
Propan-2-ol 250 mg/g
Ethanol 47 mg/g
Caution: Use biocides safely. Always read the label and product information before use.
Avantor recommend that you read the package insert and the instructions prior to each use of this product.
Documents
Specifications
- Description:Rapid Disinfectant Solution, Bacillol AF
- Seq:07
- Packed:1 litre bottle
Accessories
Product Information
- Catalog #AvailabilityPrice
- Universal sprayhead for 500 ml and 1000 ml bottles
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



