Stators, Rheodyne®
7900-183 DuraLife® II stator for MXP7970-000
Catalog # 554-2660
Supplier: IDEX RHEODYNE
Jump to:
Product Details & Documents
Stators are available in 316 stainless steel, PEEK and proprietary materials. Rheodyne® stator materials have been researched and selected for their physical and mechanical strengths. Stators only need replacing if the ports or sealing surfaces become damaged.
DuraLife® stators are stainless steel stators with the wear surface coated to prolong valve life.
DuraLife® II stators are titanium base metal with coated wear surfaces.
Specifications
- Description:7900-183 DuraLife® II stator for MXP7970-000
- Seq:04