NBVE 60/30 NL Germicidal lamp - UV-C flow chamber with 2 tubes TUV 30W and external UV-C bulb TUV 30, wall mounted, working time counter The dual-function device type NBVE 60/30 NL includes: - shielded flow UV-C chamber with 2 x TUV 30 W tubes suita 1 * 1 ST
Catalog # IBSANBVE60/30NL
Supplier: IBS Stock Items
Frequently Bought Together