QuEChERS extraction products
Tube with 900 mg anhydrous magnesium sulphate, 300 mg PSA, 150 mg graphitised carbon, 15 ml
Catalog # HYPE60105-305
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
The QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for the extraction of pesticide residues in fruit, vegetables and other foods. The QuEChERS method offers the advantage of high recoveries, accurate results and high sample throughput, combined with the associated cost savings with low solvent and labour requirements. Yet the procedure is robust and rugged.
- Simple manual procedure
- Determine wide range of pesticide types
- Less solvent usage than conventional techniques
- High recoveries, accurate results and high sample throughput
Specifications
- Capacity:15 ml
- Description:Tube with 900 mg anhydrous magnesium sulphate, 300 mg PSA, 150 mg graphitised carbon
- Seq:35