Order Entry
Slovenia
ContactUsLinkComponent
Jetpei-hepatocyte is a dna transfection reagent dedicated to the transfection of hepatic primary cells and cell lines which express galactose receptors. 1 * 100 µl
Catalog # 102-01N
undefined
Jetpei-hepatocyte is a dna transfection reagent dedicated to the transfection of hepatic primary cells and cell lines which express galactose receptors. 1 * 100 µl
Catalog # 102-01N