Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Cyclohexanone, Sigma-Aldrich®

Glass bottle, 1 L
Catalog # SIAL29140-1L
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   Merck
Cyclohexanone, Sigma-Aldrich®
Catalog # SIAL29140-1L
Supplier:  Merck
   Safer Human and Environmental Health
CAS Number:  108-94-1
Cyclohexanone, Sigma-Aldrich®
Cyclohexanone, Sigma-Aldrich®

Glass bottle, 1 L

CAS Number:  108-94-1
Price
98,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • UN:1915
  • ADR:3,III
  • REACH:01-2119453616-35

 

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Cat. No.:
    SIAL29140-1L
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications