Cyclohexanone, Sigma-Aldrich®
Glass bottle, 100 mL
Supplier: Merck
Cyclohexanone, Sigma-Aldrich®
Catalog # SIAL29140-100ML
Supplier: Merck
CAS Number: 108-94-1
Jump to:
Product Details & Documents
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Molecular Weight:98.14 g/mol
- Merck Index:13,02755
- Boiling Pt:155.6 °C (1013 hPa)
- Melting Pt:–31 °C
- Density:0.9478 g/cm³ (20 °C)
- Flash Pt:43 °C
- Storage Temperature:Ambient
- EINECS:203-631-1
- UN:1915
- ADR:3,III
- REACH:01-2119453616-35
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Cat. No.:SIAL29140-100ML
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Safer Human and Environmental Health



