Shodex MSpak PK-4A 2p - polymer based HPLC cartridge - Column Switching mode - ID: 4mm length: 10mm - Functional Group: - - Base M aterial: copolymer with N-vinyl acteoamide - Particle Size (µm): 30 - Pore Size (A): 30 1 * 1 items
Catalog # SHOWF8700012
Supplier: Resonac
Frequently Bought Together