Blue Dextran loading dye
1,5 ml, 1
Catalog # 786-556
Supplier: G-Biosciences
Jump to:
Product Details & Documents
A proprietary mixture of Formaldehyde, EDTA and Blue Dextran that is designed for use in loading DNA samples onto denaturing polyacrylamide gels.
Commonly used in loading DNA samples onto denaturing polyacrylamide gels.
Synonyms: Dextran, blue
- CAS Number:87915-38-6
- Formula:(C₂₉H₂₁N₇O₁₂S₃·3Na)n
- MDL Number:MFCD00130661
Specifications
Specifications
- Pk:1
- Pk Info:1,5 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



