Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Handheld spectroscopes

Handheld spectroscopes

Handheld spectroscope, 1501
Catalog # KRUE1501
Supplier:   A.KRÜSS Optronic
Handheld spectroscopes
Catalog # KRUE1501
Supplier:  A.KRÜSS Optronic
Handheld spectroscopes
Handheld spectroscopes

Handheld spectroscope, 1501

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

These handheld spectroscopes are designed for checking the line spectrum of gas discharge lamps as well as individual and comparative observations and spectral examinations.

  • Variable slit and angular dispersion (C-F = 7°)
  • Linear disperson of 60 mm for both models
  • Manual option with grating

Used in education for qualitative analysis and measurement of emission and absorption spectra.

Specifications
  • Model:
    1501
  • Description:
    Handheld spectroscope
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Scale illumination
    Scale illumination
    KRUE15081
    Log in for availability
    1 * 1 items - 156,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Stand for handheld spectroscopes
    Stand for handheld spectroscopes
    KRUE1510
    Log in for availability
    1 * 1 items - 165,00 €