Order Entry
Slovenia
ContactUsLinkComponent
Refrigeration bath circulator steel, sc150-a40b, -28 to +150 degree c, 1000 w cooling capacity at 20c, 6-12 l bath volume, 2.0 kw h eater, pump 17 l/min & 300 mbar usb ul/csa, 3 years warranty 230v/50hz 1 * 1 items
Catalog # HAAK153-4401
Supplier:  HAAKE
Refrigeration bath circulator steel, sc150-a40b, -28 to +150 degree c, 1000 w cooling capacity at 20c, 6-12 l bath volume, 2.0 kw h eater, pump 17 l/min & 300 mbar usb ul/csa, 3 years warranty 230v/50hz 1 * 1 items
Catalog # HAAK153-4401
Supplier:  HAAKE
Frequently Bought Together