CONNECTION FOR SUPPRESSOR ROTOR 1 * 1 items
Catalog # METR6.2832.010
Supplier: METROHM
Frequently Bought Together
Accessories for Multipette® M4
Supplier: EPPENDORF
Accessories for Transferpette® electronic pipettes
Supplier: Brand
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Accessories for HandyStep® electronic
Supplier: Brand
Accessories for vario pumps
Supplier: VACUUBRAND
Ptfe Tube 3*0,5, 600mm 1 * 1 items
Supplier: BUCHI
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
CLAMP ASSY. E200 1 * 1 items
Supplier: LAUDA
Splash protector glass 1 * 1 items
Supplier: BUCHI
Screw Cap SVL 22 1 * 1 items
Supplier: BUCHI
CAROUSEL KIT, METAL FOR 10 BOTTLES Ø35MM 1 * 1 items
Supplier: UVP ULTRA VIOLET PRODUCTS
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach
LOWER SHAFT END 1 * 1 items
Supplier: Brand