Hexanal, Sigma-Aldrich®
Glass bottle for solids, 25 g
Catalog # SIALW255726-25G-K
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Hexylaldehyde, Caproaldehyde, Hexaldehyde, Aldehyde C6, Hexyl aldehyde
- CAS Number:66-25-1
- Formula:C₆H₁₂O
- MDL Number:MFCD00007027
- Molecular Weight:100.16 g/mol
- Merck Index:13,01766
- Boiling Pt:131 °C (1013 hPa)
- Melting Pt:–56 °C
- Density:0.81 g/cm³ (20 °C)
- Flash Pt:25 °C
- Storage Temperature:Refrigerator
- UN:1207
- ADR:3,III
- :M-Clarity™ quality level = MQ100
- EINECS:200-624-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



