Hexanal, Sigma-Aldrich®
Glass bottle for solids, 1 kg
SIALW255726-1KG-K
: Merck
: Hexylaldehyde, Caproaldehyde, Hexaldehyde, Aldehyde C6, Hexyl aldehyde
- :66-25-1
- :C₆H₁₂O
- :MFCD00007027
- :100.16 g/mol
- :13,01766
- :131 °C (1013 hPa)
- :–56 °C
- :0.81 g/cm³ (20 °C)
- :25 °C
- :Refrigerator
- :1207
- :3,III
- :M-Clarity™ quality level = MQ100
- :200-624-5
- Pack type:Glass bottle for solids
- Pk:1 kg



