Order Entry
Slovenia
Orders LinkContactUsLinkComponent
1,2,6-Hexanetriol, Sigma-Aldrich®

1,2,6-Hexanetriol, Sigma-Aldrich®

Glass bottle for solids, 25 g
Catalog # SIALT66206-25G
Supplier:   Merck
1,2,6-Hexanetriol, Sigma-Aldrich®
Catalog # SIALT66206-25G
Supplier:  Merck
CAS Number:  106-69-4
1,2,6-Hexanetriol, Sigma-Aldrich®

Glass bottle for solids, 25 g

CAS Number:  106-69-4
Price
61,50 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1,2,6-Trihydroxyhexane
  • CAS Number:106-69-4
  • Formula:C₆H₁₄O₃
  • MDL Number:MFCD00002976
  • Molecular Weight:134.18 g/mol
  • Boiling Pt:323 °C (1013 hPa)
  • Melting Pt:25…32 °C
  • Density:1.11 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:203-424-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications