Order Entry
Slovenia
Orders LinkContactUsLinkComponent
1,2,6-Hexanetriol, Sigma-Aldrich®

1,2,6-Hexanetriol, Sigma-Aldrich®

Glass bottle for solids, 100 g
Catalog # SIALT66206-100G
Supplier:   Merck
1,2,6-Hexanetriol, Sigma-Aldrich®
Catalog # SIALT66206-100G
Supplier:  Merck
CAS Number:  106-69-4
1,2,6-Hexanetriol, Sigma-Aldrich®

Glass bottle for solids, 100 g

CAS Number:  106-69-4
Price
170,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,2,6-Trihydroxyhexane
  • CAS Number:106-69-4
  • Formula:C₆H₁₄O₃
  • MDL Number:MFCD00002976
  • Molecular Weight:134.18 g/mol
  • Boiling Pt:323 °C (1013 hPa)
  • Melting Pt:25…32 °C
  • Density:1.11 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:203-424-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.