Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Ethylhexanal, Sigma-Aldrich®

2-Ethylhexanal, Sigma-Aldrich®

Glass bottle, 5 mL
Catalog # SIALE29109-5ML
Supplier:   Merck
2-Ethylhexanal, Sigma-Aldrich®
Catalog # SIALE29109-5ML
Supplier:  Merck
CAS Number:  123-05-7
2-Ethylhexanal, Sigma-Aldrich®

Glass bottle, 5 mL

CAS Number:  123-05-7
Price
46,20 €
Log in for availability
1 * 5 mL
Product Details & Documents
  • CAS Number:123-05-7
  • Formula:C₈H₁₆O
  • MDL Number:MFCD00006987
  • Molecular Weight:128.21 g/mol
  • Boiling Pt:163 °C (1013 hPa)
  • Melting Pt:–67 °C
  • Density:0.82 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-596-5
  • UN:1989
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications