2-Ethylhexanal, Sigma-Aldrich®
Glass bottle, 250 mL
SIALE29109-250ML
: Merck
- :123-05-7
- :C₈H₁₆O
- :MFCD00006987
- :128.21 g/mol
- :163 °C (1013 hPa)
- :–67 °C
- :0.82 g/cm³ (20 °C)
- :Ambient
- :204-596-5
- :1989
- :3,III
- :M-Clarity™ quality level = MQ100
- Pack type:Glass bottle
- Pk:250 mL