MEGACELL DULBECCO'S MODIFIED EAGLE'S 1 * 500 mL
Catalog # SIALM3942-500ML
Supplier: Merck
Frequently Bought Together
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® M40, Transport Swab
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF