DIMETHYLOCTADECYL(3-(TRIMETHOXYSILYL)PROPYL)AMMONIUM CHLORIDE SOLUTION, 42 WT. % IN METHANOL 1 * 100 mL
SIAL435694-100ML
: Merck
DIMETHYLOCTADECYL(3-(TRIMETHOXYSILYL)PROPYL)AMMONIUM CHLORIDE SOLUTION, 42 WT. % IN METHANOL 1 * 100 mL
SIAL435694-100ML
: Merck
Frequently Bought Together
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
Sodium aluminate, Sigma-Aldrich®
: Merck