Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Starch (from corn), Sigma-Aldrich®

Starch (from corn), Sigma-Aldrich®

Glass bottle for solids, 5 kg
Catalog # SIALS4126-5KG
Supplier:   Merck
Starch (from corn), Sigma-Aldrich®
Catalog # SIALS4126-5KG
Supplier:  Merck
CAS Number:  9005-25-8
Starch (from corn), Sigma-Aldrich®

Glass bottle for solids, 5 kg

CAS Number:  9005-25-8
Price
231,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
  • CAS Number:9005-25-8
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00082026
  • Molecular Weight:162.14 g/mol
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:232-679-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.