Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Starch (from corn), Sigma-Aldrich®

Glass bottle for solids, 2 kg
Catalog # SIALS4126-2KG
Supplier:   Merck
Starch (from corn), Sigma-Aldrich®
Catalog # SIALS4126-2KG
Supplier:  Merck
CAS Number:  9005-25-8
Starch (from corn), Sigma-Aldrich®

Glass bottle for solids, 2 kg

CAS Number:  9005-25-8
Price
86,00 €
Log in for availability
1 * 2 kg
Product Details & Documents
  • CAS Number:9005-25-8
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00082026
  • Molecular Weight:162.14 g/mol
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:232-679-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2 kg

Specifications