D-(-)-Mannitol, Sigma-Aldrich®
Glass bottle for solids, 100 g
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Merck Index:13,05769
- Boiling Pt:290…295 °C (4 hPa)
- Melting Pt:164…169 °C
- Density:1.49 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:200-711-8
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



