Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Cyclohexanone, Sigma-Aldrich®

Cyclohexanone, Sigma-Aldrich®

Glass bottle, 1 L
Catalog # SIALC102180-1L
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   Merck
Cyclohexanone, Sigma-Aldrich®
Catalog # SIALC102180-1L
Supplier:  Merck
   Safer Human and Environmental Health
CAS Number:  108-94-1
Cyclohexanone, Sigma-Aldrich®
Cyclohexanone, Sigma-Aldrich®

Glass bottle, 1 L

CAS Number:  108-94-1
Price
63,20 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • UN:1915
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • REACH:01-2119453616-35

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Cat. No.:
    SIALC102180-1L
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.