Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

2-Fluorocyclohexanone 96%

Glass bottle for solids, 1 g
Catalog # H26032.03
Irritant
2-Fluorocyclohexanone 96%
Catalog # H26032.03
CAS Number:  694-82-6
2-Fluorocyclohexanone 96%
2-Fluorocyclohexanone 96%

Glass bottle for solids, 1 g

CAS Number:  694-82-6
Price
230,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 96% CAS No.: 694-82-6 Molecular Formula: C6H9FO Formula Weight: 116.13 Storage Temperature: -30°C to -10°C Flash Point: 71°(160°F) Refractive Index: 1.4440 Boiling Point: 78°/17mm MDL No.: MFCD01076476

  • CAS Number:694-82-6
  • Formula:C₆H₉FO
  • MDL Number:MFCD01076476
  • Molecular Weight:116.14 g/mol
  • Boiling Pt:78 °C (17 mmHg)
  • Density:1.08
  • Flash Pt:71 °C (160 °F)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.