Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Fluorocyclohexanone 96%

2-Fluorocyclohexanone 96%

Glass bottle for solids, 250 mg
Catalog # H26032.MD
Irritant
2-Fluorocyclohexanone 96%
Catalog # H26032.MD
CAS Number:  694-82-6
2-Fluorocyclohexanone 96%
2-Fluorocyclohexanone 96%

Glass bottle for solids, 250 mg

CAS Number:  694-82-6
Price
86,70 €
Log in for availability
1 * 250 mg
Product Details & Documents

Purity: 96% CAS No.: 694-82-6 Molecular Formula: C6H9FO Formula Weight: 116.13 Storage Temperature: -30°C to -10°C Flash Point: 71°(160°F) Refractive Index: 1.4440 Boiling Point: 78°/17mm MDL No.: MFCD01076476

  • CAS Number:694-82-6
  • Formula:C₆H₉FO
  • MDL Number:MFCD01076476
  • Molecular Weight:116.14 g/mol
  • Boiling Pt:78 °C (17 mmHg)
  • Density:1.08
  • Flash Pt:71 °C (160 °F)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg

Specifications