HEPES BIOXTRA, PH 5.0-6.5 (20 C, 1 M IN H2O) 1 * 250 g
Catalog # SIALH7523-250G
Supplier: Merck
Frequently Bought Together
Ethylenediamine ≥99.0% (by GC), GPR RECTAPUR®
Supplier: VWR Chemicals
Water, Ultrapure for HPLC
Supplier: Thermo Fisher Scientific
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, anhydrous 99.9% stabilised, extra pure
Supplier: Thermo Fisher Scientific
n-Heptane, anhydrous, over molecular sieves
Supplier: Thermo Fisher Scientific
POTASSIUM NITRATE-15N, 98 ATOM % 15N 1 * 1 items
Supplier: Merck
Acetonitrile, extra dry 99.9+%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals