Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, suitable only for use with dual block model, 96 well plate, skirted or non-skirted or 4 slides, —
Catalog # JENC357-138
Supplier: JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Catalog # JENC357-138
Supplier: JENCONS SCIENTIFIC
Jump to:
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Specifications
- No. of Holes:—
- Description:Interchangeable block, suitable only for use with dual block model
- Seq:17
- For:96 well plate, skirted or non-skirted or 4 slides